accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHHO

CAS RN 2896-60-8

IUPAC Name: 4-ethylbenzene-1,3-diol
Synonyms: 1,3-benzenediol, 4-ethyl-
Molecular Weight (g/mol): 138.17
Molecular Formula: C8H10O2
InChi Key: VGMJYYDKPUPTID-UHFFFAOYSA-N
SMILES: CCC1=CC=C(O)C=C1O
Molecular Weight (g/mol) 
  • (1)
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (1)
  • (1)
  • (3)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (1)
  • (5)
  • (2)
  • (2)
  • (6)
  • (1)
  • (1)
  • (3)
  • (3)
  • (4)
  • (2)