accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaOSOOHNHNSOOONSOOO

CAS RN 28983-56-4

IUPAC Name: disodium 2-{[4-({4-[(2-sulfonatophenyl)amino]phenyl}({4-[(2-sulfonatophenyl)imino]cyclohexa-2,5-dien-1-ylidene})methyl)phenyl]amino}benzene-1-sulfonate
Synonyms: methyl blue
Molecular Weight (g/mol): 798.79
Molecular Formula: C37H26N3Na2O9S3
InChi Key: TUHAIJABPUJAEY-UHFFFAOYSA-K
SMILES: [Na+].[Na+].[O-]S(=O)(=O)C1=CC=CC=C1NC1=CC=C(C=C1)C(C1=CC=C(NC2=CC=CC=C2S([O-])(=O)=O)C=C1)=C1C=CC(C=C1)=NC1=CC=CC=C1S([O-])(=O)=O
Color 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results