accessibility menu, dialog, popup

UserName

Viewing profile as user:
NiSSSS

CAS RN 28984-20-5

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 543.4
Molecular Formula: C28H20NiS4
InChi Key: LJISAPFYPQSNRX-DEWSNNOYSA-J
SMILES: [Ni].[S-]\C(=C(/[S-])C1=CC=CC=C1)C1=CC=CC=C1.[S-]\C(=C(/[S-])C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results