accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNN

CAS RN 290-87-9

IUPAC Name: 1,3,5-triazine
Synonyms: 1,3,5-triazine vedita 250
Molecular Weight (g/mol): 81.08
Molecular Formula: C3H3N3
InChi Key: JIHQDMXYYFUGFV-UHFFFAOYSA-N
SMILES: C1=NC=NC=N1
Percent Purity 
  • (3)
  • (5)
Quantity 
  • (3)
  • (2)
  • (3)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (3)
  • (5)
  • (3)
  • (2)
  • (3)
  • (6)
  • (3)
  • (3)