accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHNOHO

CAS RN 2901-75-9

IUPAC Name: 2-acetamido-3-phenylpropanoic acid
Synonyms: N-acetyl-D,L-phenylalanine afalanine afalanina afalaninum N-acetyl-3-phenyl-DL-alanine DL-N-acetyl-3-phenylalanine
Molecular Weight (g/mol): 207.23
Molecular Formula: C11H13NO3
InChi Key: CBQJSKKFNMDLON-UHFFFAOYNA-N
SMILES: CC(=O)NC(CC1=CC=CC=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results