accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 29027-20-1

IUPAC Name: 3-chloro-5-methylaniline
Synonyms: 3-chloro-5-methylaniline
Molecular Weight (g/mol): 141.6
Molecular Formula: C7H8ClN
InChi Key: XUZCJDBXXYJXDD-UHFFFAOYSA-N
SMILES: CC1=CC(N)=CC(Cl)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results