accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 290331-71-4

IUPAC Name: {1-[(tert-butoxy)carbonyl]-5-methoxy-1H-indol-2-yl}boronic acid
Synonyms: 1-(tert-butoxycarbonyl)-5-methoxyindol-2-ylboronic acid
Molecular Weight (g/mol): 291.11
Molecular Formula: C14H18BNO5
InChi Key: PZLVPMBCKHDVKT-UHFFFAOYSA-N
SMILES: COC1=CC=C2N(C(=O)OC(C)(C)C)C(=CC2=C1)B(O)O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)