accessibility menu, dialog, popup

UserName

Viewing profile as user:
SOOClBr

CAS RN 2905-25-1

IUPAC Name: 2-bromobenzene-1-sulfonyl chloride
Synonyms: 2-bromobenzenesulfonyl chloride
Molecular Weight (g/mol): 255.51
Molecular Formula: C6H4BrClO2S
InChi Key: VFPWGZNNRSQPBT-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CC=CC=C1Br
Molecular Weight (g/mol) 
  • (6)
  • (1)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (3)
Boiling Point 
  • (2)
  • (2)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (6)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (4)