accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCl

CAS RN 2905-65-9

IUPAC Name: methyl 3-chlorobenzoate
Synonyms: methyl 3-chlorobenzoate
Molecular Weight (g/mol): 170.59
Molecular Formula: C8H7ClO2
InChi Key: XRDRKVPNHIWTBX-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC(Cl)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results