CAS RN 2905-68-2
IUPAC Name:
methyl 3,4-dichlorobenzoate
Synonyms:
methyl 3,4-dichlorobenzoate
Molecular Weight (g/mol):
205.03
Molecular Formula:
C8H6Cl2O2
InChi Key:
WICGATIORFSOJL-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC=C(Cl)C(Cl)=C1
1
–
1
of
1
results
1
Promotions Available
Methyl 3,4-dichlorobenzoate, 97%, Thermo Scientific Chemicals