accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2906-60-7

IUPAC Name: 4-[(3-carboxylatopropyl)disulfanyl]butanoate
Synonyms: 4-[(3-carboxylatopropyl)disulfanyl]butanoate
Molecular Weight (g/mol): 236.3
Molecular Formula: C8H12O4S2
InChi Key: YYSCJLLOWOUSHH-UHFFFAOYSA-L
SMILES: [O-]C(=O)CCCSSCCCC([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)