accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 29085-13-0

IUPAC Name: dimethyl[(methylsulfanyl)methylidene]azanium iodide
Synonyms: dimethyl[(methylsulfanyl)methylidene]azanium iodide
Molecular Weight (g/mol): 231.1
Molecular Formula: C4H10INS
InChi Key: BPBAGFOLLWBKGJ-UHFFFAOYSA-M
SMILES: [I-].CSC=[N+](C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results