accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 29114-66-7

IUPAC Name: 1-(4-fluorophenyl)pentan-1-one
Synonyms: 1-(4-fluorophenyl)pentan-1-one
Molecular Weight (g/mol): 180.22
Molecular Formula: C11H13FO
InChi Key: HBJRRAMFTUDWMQ-UHFFFAOYSA-N
SMILES: CCCCC(=O)C1=CC=C(F)C=C1
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)