accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C

CAS RN 29136-19-4

IUPAC Name: nonadecylbenzene
Synonyms: benzene, nonadecyl-
Molecular Weight (g/mol): 344.63
Molecular Formula: C25H44
InChi Key: SHWJJBRTHGGZBE-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results