accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2917-46-6

IUPAC Name: bis(chloromethyl)dimethylsilane
Synonyms: bis(chloromethyl)dimethylsilane
Molecular Weight (g/mol): 157.11
Molecular Formula: C4H10Cl2Si
InChi Key: TVRFAOJPBXYIRM-UHFFFAOYSA-N
SMILES: C[Si](C)(CCl)CCl
Boiling Point 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)