accessibility menu, dialog, popup

UserName

Viewing profile as user:
NClNCl

CAS RN 29176-55-4

IUPAC Name: 2,9-dichloro-1,10-phenanthroline
Synonyms: 2,9-dichloro-1,10-phenanthroline
Molecular Weight (g/mol): 249.09
Molecular Formula: C12H6Cl2N2
InChi Key: DNKGIDURJINUOA-UHFFFAOYSA-N
SMILES: ClC1=CC=C2C=CC3=CC=C(Cl)N=C3C2=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results