accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNN

CAS RN 29181-50-8

IUPAC Name: pyridine-2,4-dicarbonitrile
Synonyms: pyridine-2,4-dicarbonitrile
Molecular Weight (g/mol): 129.12
Molecular Formula: C7H3N3
InChi Key: HLAGQMFURMNTLW-UHFFFAOYSA-N
SMILES: N#CC1=CC=NC(=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results