accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 29199-09-5

IUPAC Name: 3',6'-bis(diethylamino)-2-(4-nitrophenyl)-2,3-dihydrospiro[isoindole-1,9'-xanthen]-3-one
Synonyms: 3',6'-bis(diethylamino)-2-(4-nitrophenyl)spiro[isoindole-1,9'-xanthen]-3-one
Molecular Weight (g/mol): 562.67
Molecular Formula: C34H34N4O4
InChi Key: XZXFZILEZWXEND-UHFFFAOYSA-N
SMILES: CCN(CC)C1=CC=C2C(OC3=CC(=CC=C3C22N(C(=O)C3=CC=CC=C23)C2=CC=C(C=C2)[N+]([O-])=O)N(CC)CC)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results