CAS RN 29211-70-9
IUPAC Name:
3-propyl-4,5-dihydro-1H-pyrazol-5-one
Synonyms:
5-propyl-2,4-dihydropyrazol-3-one
Molecular Weight (g/mol):
126.16
Molecular Formula:
C6H10N2O
InChi Key:
GVUNLYBSNQOHBD-UHFFFAOYSA-N
SMILES:
CCCC1=NNC(=O)C1
1
–
1
of
1
results
1
Promotions Available
3-n-Propyl-2-pyrazolin-5-one, 98%
3-n-Propyl-2-pyrazolin-5-one, 98%
CAS: 29211-70-9 Molecular Formula: C6H10N2O Molecular Weight (g/mol): 126.16 MDL Number: MFCD00051646 InChI Key: GVUNLYBSNQOHBD-UHFFFAOYSA-N PubChem CID: 269244 IUPAC Name: 3-propyl-1,4-dihydropyrazol-5-one SMILES: CCCC1=NNC(=O)C1