accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 29214-60-6

IUPAC Name: ethyl 2-acetyloctanoate
Synonyms: ethyl 2-acetyloctanoate
Molecular Weight (g/mol): 214.31
Molecular Formula: C12H22O3
InChi Key: BRGRZPQESQKATK-UHFFFAOYNA-N
SMILES: CCCCCCC(C(C)=O)C(=O)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results