accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2922-54-5

IUPAC Name: sodium 3-carboxypropanoate
Synonyms: sodium succinate
Molecular Weight (g/mol): 140.07
Molecular Formula: C4H5NaO4
InChi Key: KZQSXALQTHVPDQ-UHFFFAOYSA-M
SMILES: [Na+].OC(=O)CCC([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results