accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFHNNH2HCl

CAS RN 2923-56-0

IUPAC Name: [4-(trifluoromethyl)phenyl]hydrazine hydrochloride
Synonyms: [4-(trifluoromethyl)phenyl]hydrazine hydrochloride
Molecular Weight (g/mol): 212.6
Molecular Formula: C7H8ClF3N2
InChi Key: WCAGNYIHAYOPSE-UHFFFAOYSA-N
SMILES: Cl.NNC1=CC=C(C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results