accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrH2NF

CAS RN 2924-09-6

IUPAC Name: 5-bromo-2-fluoroaniline
Synonyms: 5-bromo-2-fluoroaniline
Molecular Weight (g/mol): 190.02
Molecular Formula: C6H5BrFN
InChi Key: ADWKOCXRCRSMLQ-UHFFFAOYSA-N
SMILES: NC1=C(F)C=CC(Br)=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)