accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 292633-16-0

H3COOHNOOHONHONHNHNOOHONHOH3CCH3HNOCH3

CAS RN 292633-16-0

IUPAC Name: 4-{[1-({2-carboxy-1-[(4-methyl-2-oxo-2H-chromen-7-yl)carbamoyl]ethyl}carbamoyl)-2-(1H-imidazol-5-yl)ethyl]carbamoyl}-4-(2-acetamido-4-methylpentanamido)butanoic acid
Synonyms: 4-{[1-({2-carboxy-1-[(4-methyl-2-oxochromen-7-yl)carbamoyl]ethyl}carbamoyl)-2-(3H-imidazol-4-yl)ethyl]carbamoyl}-4-(2-acetamido-4-methylpentanamido)butanoic acid
Molecular Weight (g/mol): 711.73
Molecular Formula: C33H41N7O11
InChi Key: ZILMZAQLFLKJPG-UHFFFAOYNA-N
SMILES: CC(C)CC(NC(C)=O)C(=O)NC(CCC(O)=O)C(=O)NC(CC1=CN=CN1)C(=O)NC(CC(O)=O)C(=O)NC1=CC=C2C(C)=CC(=O)OC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)