accessibility menu, dialog, popup

UserName

Viewing profile as user:
NONF

CAS RN 29270-55-1

IUPAC Name: 4-fluoro-2,1,3-benzoxadiazole
Synonyms: 4-fluoro-2,1,3-benzoxadiazole
Molecular Weight (g/mol): 138.1
Molecular Formula: C6H3FN2O
InChi Key: QKZQJBBZKDVVFI-UHFFFAOYSA-N
SMILES: FC1=CC=CC2=NON=C12
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results