accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 29274-28-0

IUPAC Name: 4-chloro-1H-pyrazolo[3,4-b]pyridine
Synonyms: 4-chloro-1H-pyrazolo[3,4-b]pyridine
Molecular Weight (g/mol): 153.57
Molecular Formula: C6H4ClN3
InChi Key: AKWQSKVILVJWPC-UHFFFAOYSA-N
SMILES: ClC1=C2C=NNC2=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results