accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 294-62-2

IUPAC Name: cyclododecane
Synonyms: cyclododecane
Molecular Weight (g/mol): 168.32
Molecular Formula: C12H24
InChi Key: DDTBPAQBQHZRDW-UHFFFAOYSA-N
SMILES: C1CCCCCCCCCCC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results