accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2941-23-3

IUPAC Name: 2-aminocyclopent-1-ene-1-carbonitrile
Synonyms: 2-aminocyclopent-1-ene-1-carbonitrile
Molecular Weight (g/mol): 108.14
Molecular Formula: C6H8N2
InChi Key: NSMYBPIHVACKQG-UHFFFAOYSA-N
SMILES: NC1=C(CCC1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results