accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOSN

CAS RN 2942-07-6

IUPAC Name: 5-nitro-1,3-benzothiazole
Synonyms: 5-nitro-1,3-benzothiazole
Molecular Weight (g/mol): 180.18
Molecular Formula: C7H4N2O2S
InChi Key: AEUQLELVLDMMKB-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C2SC=NC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results