accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCH3H3C

CAS RN 2944-47-0

IUPAC Name: 1-methoxy-2-(propan-2-yl)benzene
Synonyms: 1-isopropyl-2-methoxybenzene
Molecular Weight (g/mol): 150.22
Molecular Formula: C10H14O
InChi Key: NNZRVXTXKISCGS-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1C(C)C
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results