accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2947-60-6

IUPAC Name: 2-(3-methylphenyl)acetonitrile
Synonyms: 1-methyl-3-cyanomethylbenzene
Molecular Weight (g/mol): 131.18
Molecular Formula: C9H9N
InChi Key: WOJADIOTNFDWNQ-UHFFFAOYSA-N
SMILES: CC1=CC=CC(CC#N)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results