accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3N

CAS RN 2947-61-7

IUPAC Name: 2-(4-methylphenyl)acetonitrile
Synonyms: tolylacetonitrile
Molecular Weight (g/mol): 131.18
Molecular Formula: C9H9N
InChi Key: RNHKXHKUKJXLAU-UHFFFAOYSA-N
SMILES: CC1=CC=C(CC#N)C=C1
Boiling Point 
  • (3)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (5)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)