accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHNHH3C

CAS RN 29475-64-7

IUPAC Name: 2-(methylamino)propanoic acid
Synonyms: 2-(methylamino)propanoic acid
Molecular Weight (g/mol): 103.12
Molecular Formula: C4H9NO2
InChi Key: GDFAOVXKHJXLEI-UHFFFAOYNA-N
SMILES: CNC(C)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)