accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 29477-27-8

IUPAC Name: N-cyclohexyl-N-methyl-4-nitrobenzamide
Synonyms: N-cyclohexyl-N-methyl-4-nitrobenzamide
Molecular Weight (g/mol): 262.31
Molecular Formula: C14H18N2O3
InChi Key: ZGDQZPUGCAXWLR-UHFFFAOYSA-N
SMILES: CN(C1CCCCC1)C(=O)C1=CC=C(C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results