CAS RN 294877-33-1
IUPAC Name:
1-(3-bromophenyl)-1H-pyrazole
Synonyms:
1-(3-bromophenyl)pyrazole
Molecular Weight (g/mol):
223.07
Molecular Formula:
C9H7BrN2
InChi Key:
KUWAIOZTKMAIPI-UHFFFAOYSA-N
SMILES:
BrC1=CC=CC(=C1)N1C=CC=N1
1
–
1
of
1
results
1
1-(3-Bromophenyl)-1H-pyrazole, ≥97%, Thermo Scientific™
CAS: 294877-33-1 Molecular Formula: C9H7BrN2 Molecular Weight (g/mol): 223.073 MDL Number: MFCD06797472 InChI Key: KUWAIOZTKMAIPI-UHFFFAOYSA-N Synonym: 1-3-bromophenyl-1h-pyrazole,1-3-bromophenyl pyrazole,1-3-bromo-phenyl-1h-pyrazole,3-1h-pyrazol-1-yl bromobenzene,3-bromophenyl pyrazole,acmc-209h9b,1h-pyrazole,1-3-bromophenyl,1h-pyrazole, 1-3-bromophenyl PubChem CID: 7127804 IUPAC Name: 1-(3-bromophenyl)pyrazole SMILES: C1=CC(=CC(=C1)Br)N2C=CC=N2