accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 295-14-7

NHNHHNHN

CAS RN 295-14-7

IUPAC Name: 1,4,7,10-tetraazacyclotridecane
Synonyms: 1,4,7,10-tetraazacyclotridecane
Molecular Weight (g/mol): 186.3
Molecular Formula: C9H22N4
InChi Key: LADZJJOUGVGJHM-UHFFFAOYSA-N
SMILES: C1CNCCNCCNCCNC1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results