accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHNHHNHN

CAS RN 295-37-4

IUPAC Name: 1,4,8,11-tetraazacyclotetradecane
Synonyms: cyclam
Molecular Weight (g/mol): 200.33
Molecular Formula: C10H24N4
InChi Key: MDAXKAUIABOHTD-UHFFFAOYSA-N
SMILES: C1CNCCNCCCNCCNC1
Molecular Weight (g/mol) 
  • (2)
  • (8)
Quantity 
  • (4)
  • (2)
  • (4)
Percent Purity 
  • (3)
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (2)
  • (6)
  • (2)
  • (8)
  • (4)
  • (2)
  • (4)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)