accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 29515-99-9

IUPAC Name: 5-(methylsulfanyl)-2H-1,2,3,4-tetrazole
Synonyms: 5-(methylsulfanyl)-2H-1,2,3,4-tetrazole
Molecular Weight (g/mol): 116.14
Molecular Formula: C2H4N4S
InChi Key: ZBXNFTFKKOSPLD-UHFFFAOYSA-N
SMILES: CSC1=NNN=N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results