accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 295349-64-3

IUPAC Name: 2-(morpholin-4-yl)pyridine-4-carboxylic acid
Synonyms: 2-(morpholin-4-yl)pyridine-4-carboxylic acid
Molecular Weight (g/mol): 208.22
Molecular Formula: C10H12N2O3
InChi Key: OVOIVNZYILYGKU-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=NC=C1)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results