accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2954-50-9

IUPAC Name: 1,2,3,4-tetrahydronaphthalen-2-amine hydrochloridyl
Synonyms: 2-aminotetralin
Molecular Weight (g/mol): 182.67
Molecular Formula: C10H13ClN
InChi Key: GIXZIEQZNPTVNL-UHFFFAOYNA-N
SMILES: [Cl].NC1CCC2=CC=CC=C2C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results