accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 29540-81-6

IUPAC Name: 1-trifluoromethanesulfonyl-1H-imidazole
Synonyms: 1-trifluoromethanesulfonylimidazole
Molecular Weight (g/mol): 200.14
Molecular Formula: C4H3F3N2O2S
InChi Key: YGABUCCNCBMODG-UHFFFAOYSA-N
SMILES: FC(F)(F)S(=O)(=O)N1C=CN=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results