accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOH3CH3CH3CH3C(E)(E)

CAS RN 29548-30-9

IUPAC Name: (2E,6E)-3,7,11-trimethyldodeca-2,6,10-trien-1-yl acetate
Synonyms: farnesyl acetate
Molecular Weight (g/mol): 264.41
Molecular Formula: C17H28O2
InChi Key: ZGIGZINMAOQWLX-NCZFFCEISA-N
SMILES: CC(C)=CCC\C(C)=C\CC\C(C)=C\COC(C)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)