accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2955-88-6

IUPAC Name: 2-(pyrrolidin-1-yl)ethan-1-ol
Synonyms: epolamine 1-pyrrolidineethanol HEP
Molecular Weight (g/mol): 115.18
Molecular Formula: C6H13NO
InChi Key: XBRDBODLCHKXHI-UHFFFAOYSA-N
SMILES: OCCN1CCCC1
Molecular Weight (g/mol) 
  • (2)
  • (7)
Percent Purity 
  • (2)
  • (3)
  • (4)
Quantity 
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (5)
  • (2)
  • (7)
  • (2)
  • (3)
  • (4)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (5)