accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 29555-02-0

IUPAC Name: (1R,2S)-2-methylcyclopropane-1-carboxylate
Synonyms: (1R,2S)-2-methylcyclopropane-1-carboxylate
Molecular Weight (g/mol): 99.11
Molecular Formula: C5H7O2
InChi Key: AYEGPMGNMOIHDL-IUYQGCFVSA-M
SMILES: C[C@H]1C[C@H]1C([O-])=O
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)