accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 29559-55-5

IUPAC Name: 1,3-dichloro-2,2-dimethylpropane
Synonyms: 1,3-dichloro-2,2-dimethylpropane
Molecular Weight (g/mol): 141.04
Molecular Formula: C5H10Cl2
InChi Key: KTWNITKLQPCZSL-UHFFFAOYSA-N
SMILES: CC(C)(CCl)CCl
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results