accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2958-62-5

IUPAC Name: 2-isocyanato-1,3,5-trimethylbenzene
Synonyms: 2-isocyanato-1,3,5-trimethylbenzene
Molecular Weight (g/mol): 161.2
Molecular Formula: C10H11NO
InChi Key: HJHZRZFONUPQAA-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(N=C=O)C(C)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results