accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(S)ClOO

CAS RN 29617-66-1

IUPAC Name: (2S)-2-chloropropanoate
Synonyms: (2S)-2-chloropropanoate
Molecular Weight (g/mol): 107.51
Molecular Formula: C3H4ClO2
InChi Key: GAWAYYRQGQZKCR-REOHCLBHSA-M
SMILES: C[C@H](Cl)C([O-])=O
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (5)
  • (4)