accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNSHNONSHN

CAS RN 29635-94-7

IUPAC Name: 3-[4-(5-oxo-1-phenyl-2-sulfanylideneimidazolidin-4-yl)butyl]-1-phenylthiourea
Synonyms: 3-[4-(5-oxo-1-phenyl-2-sulfanylideneimidazolidin-4-yl)butyl]-1-phenylthiourea
Molecular Weight (g/mol): 398.54
Molecular Formula: C20H22N4OS2
InChi Key: BUPYRERCBPFRFM-UHFFFAOYNA-N
SMILES: O=C1C(CCCCNC(=S)NC2=CC=CC=C2)NC(=S)N1C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results