accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHNHO

CAS RN 29636-87-1

IUPAC Name: (5-methyl-1H-imidazol-4-yl)methanol
Synonyms: (5-methyl-1H-imidazol-4-yl)methanol
Molecular Weight (g/mol): 112.13
Molecular Formula: C5H8N2O
InChi Key: AXJZCJSXNZZMDU-UHFFFAOYSA-N
SMILES: CC1=C(CO)N=CN1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results