CAS RN 29663-54-5
IUPAC Name:
cyclohepta-2,4,6-trien-1-ylium; hexafluoro-λ⁵-phosphanuide
Synonyms:
tropylium hexafluorophosphate
Molecular Weight (g/mol):
236.1
Molecular Formula:
C7H7F6P
InChi Key:
ZCBPSALVMPRNKG-UHFFFAOYSA-N
SMILES:
F[P-](F)(F)(F)(F)F.[CH+]1C=CC=CC=C1
1
–
1
of
1
results
1
Tropylium hexafluorophosphate, Thermo Scientific™
CAS: 29663-54-5 Molecular Formula: C7H7·PF6 MDL Number: MFCD00013403